siRNA Details
Gene Symbol: Mkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122667.2 | NP_001116139.1 | 10 | 1034 | ||
NM_153588.3 | NP_705816.2 | 8 | 1048 | |||
GenBank mRNA | AF532598.1 | - | 8 | 830 | ||
AK220377.1 | - | 8 | 830 | |||
BC131644.1 | - | 8 | 830 | 40110019 | ||
BC131645.1 | - | 8 | 828 |
