siRNA Details
Gene Symbol: Mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041672.1 | - | 1657 | |||
AK164429.1 | - | 2045 | ||||
AK171197.1 | - | 2777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
