siRNA Details
Gene Symbol: Cep164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081373.2 | NP_001074842.2 | 23, 24 | 3083 | ||
GenBank mRNA | AK050991.1 | - | 2, 3 | 168 | ||
AK220365.1 | - | 19, 20 | 2426 | |||
BC066145.1 | - | 5, 6 | 525 | |||
BC076567.1 | - | 21, 22 | 2824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
