siRNA Details
Gene Symbol: Cep164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081373.2 | NP_001074842.2 | 30 | 5464 | ||
GenBank mRNA | AK050991.1 | - | 9 | 2550 | ||
AK220365.1 | - | 26 | 4808 | |||
BB206864.2 | - | 1 | 645 | |||
BC066145.1 | - | 12 | 2905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
