siRNA Details
Gene Symbol: Ganc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172672.2 | NP_766260.2 | 3, 4 | 1336 | ||
GenBank mRNA | AK034155.1 | - | 3, 4 | 470 | ||
AK036238.1 | - | 3, 4 | 1337 | |||
AK076333.1 | - | 2, 3 | 325 | |||
AK135218.1 | - | 3, 4 | 1020 | |||
AK220303.1 | - | 3, 4 | 1356 | |||
BC075687.1 | - | 3, 4 | 607 | |||
BC079548.1 | - | 3, 4 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
