siRNA Details
Gene Symbol: Ganc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172672.2 | NP_766260.2 | 5 | 1546 | ||
GenBank mRNA | AK034155.1 | - | 5 | 680 | ||
AK036238.1 | - | 5 | 1547 | |||
AK076333.1 | - | 4 | 490 | |||
AK135218.1 | - | 5 | 1230 | |||
AK220303.1 | - | 5 | 1566 | |||
BC075687.1 | - | 5 | 817 | |||
BC079548.1 | - | 5 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
