siRNA Details
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081238.2 | NP_001074707.1 | 4 | 796 | ||
NM_001289842.1 | NP_001276771.1 | 4 | 796 | |||
NM_001289843.1 | NP_001276772.1 | 3 | 702 | |||
GenBank mRNA | AK028653.1 | - | 4 | 770 | ||
AK154227.1 | - | 4 | 768 | |||
AK156701.1 | - | 4 | 715 | |||
AK157382.1 | - | 4 | 715 | |||
BC066139.1 | - | 3 | 644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
