siRNA Details
Gene Symbol: Loxhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172834.2 | NP_766422.2 | 7, 8 | 891 | ||
GenBank mRNA | AK030047.1 | - | 4, 5 | 726 | ||
FJ750876.1 | - | 7, 8 | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
