siRNA Details
Gene Symbol: BMP2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017593.3 | NP_060063.2 | 14 | 2142 | ||
GenBank mRNA | AF527532.1 | - | 2 | 654 | ||
AK075278.1 | - | 10 | 1337 | |||
AK292147.1 | - | 14 | 2118 | |||
AL137661.1 | - | 16 | 2174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
