siRNA Details
Gene Symbol: TRPM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301212.1 | NP_001288141.1 | 39 | 6024 | ||
NM_017672.5 | NP_060142.3 | 39 | 6027 | |||
XM_005254486.3 | XP_005254543.1 | 40 | 6082 | |||
XM_017022350.1 | XP_016877839.1 | 40 | 5847 | |||
XM_017022351.1 | XP_016877840.1 | 40 | 5844 | |||
XM_017022352.1 | XP_016877841.1 | 39 | 5787 | |||
XM_017022353.1 | XP_016877842.1 | 39 | 5784 | |||
XM_017022354.1 | XP_016877843.1 | 40 | 5912 | |||
XM_017022355.1 | XP_016877844.1 | 38 | 5714 | |||
GenBank mRNA | AF346629.2 | - | 39 | 5980 | ||
AK075193.1 | - | 5 | 732 | |||
AK172800.1 | - | 16 | 2472 | |||
AY032950.1 | - | 39 | 6016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
