siRNA Details
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172887.2 | NP_766475.2 | 4 | 764 | ||
GenBank mRNA | AK086708.1 | - | 4 | 652 | ||
AK157981.1 | - | 4 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
