siRNA Details
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175561.4 | NP_780770.2 | 5 | 1339 | ||
GenBank mRNA | AK043903.1 | - | 3 | 700 | ||
BC068235.1 | - | 5 | 1315 | |||
BC132334.1 | - | 1 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
