siRNA Details
Gene Symbol: Gm6182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082611.1 | - | 887 | |||
AK165101.1 | - | 889 | ||||
BC080301.1 | - | 859 | ||||
BC090643.1 | - | 591 |
Gene Symbol: BC048559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133171.1 | - | 931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
