siRNA Details
Gene Symbol: 9930111J21Rik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173434.1 | NP_775610.1 | 3 | 789 | ||
GenBank mRNA | AK037088.1 | - | 3 | 789 | ||
BC066104.1 | - | 777 | ||||
BC119805.1 | - | 2 | 693 | 40107440 | ||
BC122882.1 | - | 2 | 698 | 40107441 |
Gene Symbol: 9930111J21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114679.1 | NP_001108151.1 | 776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
