siRNA Details
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282647.1 | NP_001269576.1 | 5 | 1071 | ||
NM_005660.2 | NP_005651.1 | 5 | 1661 | |||
GenBank mRNA | D88146.1 | - | 5 | 1343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
