siRNA Details
Gene Symbol: E230001N04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053925.1 | - | 1 | 1094 | ||
AK153588.1 | - | 1 | 1081 | |||
AK162363.1 | - | 1 | 746 | |||
BC096647.1 | - | 1 | 1091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
