siRNA Details
Gene Symbol: C130071C03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029460.1 | - | 2229 | |||
AK048540.1 | - | 2230 | ||||
KF032589.1 | - | 3284 | ||||
KF032590.1 | - | 3209 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C130071C03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029460.1 | - | 2229 | |||
AK048540.1 | - | 2230 | ||||
KF032589.1 | - | 3284 | ||||
KF032590.1 | - | 3209 |