siRNA Details
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029471.1 | - | 2 | 857 | ||
AK077292.1 | - | 2 | 804 | |||
AK160463.1 | - | 3 | 1005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
