siRNA Details
Gene Symbol: Maats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081025.1 | NP_001074494.1 | 2 | 266 | ||
GenBank mRNA | AF503943.1 | - | 2 | 549 | ||
AK045121.1 | - | 2 | 218 | |||
AK077038.1 | - | 2 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
