siRNA Details
Gene Symbol: Dlec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177117.3 | NP_796091.2 | 3 | 638 | ||
GenBank mRNA | AK045848.1 | - | 3 | 639 | ||
AK133987.1 | - | 3 | 609 | |||
BC145277.1 | - | 3 | 621 | |||
BC145278.1 | - | 3 | 621 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
