siRNA Details
Gene Symbol: Kirrel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170985.1 | NP_001164456.1 | 16 | 6701 | ||
NM_130867.3 | NP_570937.2 | 16 | 6698 | |||
GenBank mRNA | AK032898.1 | - | 1 | 3280 | ||
AK034569.1 | - | 1 | 1910 | |||
BC062651.1 | - | 1 | 1743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
