siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039376.2 | NP_001034465.2 | 4 | 498 | ||
NM_001289701.1 | NP_001276630.1 | 4 | 498 | |||
NM_001289702.1 | NP_001276631.1 | 4 | 498 | |||
GenBank mRNA | AK031066.1 | - | 1 | 266 | ||
AK051209.1 | - | 4 | 576 | |||
AK083867.1 | - | 4 | 573 | |||
AK141917.1 | - | 4 | 624 | |||
BC028274.1 | - | 4 | 508 | |||
BC141185.1 | - | 4 | 498 | 9055797 | ||
BC145504.1 | - | 4 | 498 | 9054067 | ||
CN695623.1 | - | 1 | 367 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_022382, AF139185.
Gene Ontology Categories:
