siRNA Details
Gene Symbol: Kcnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008421.3 | NP_032447.1 | 2 | 7504 | ||
GenBank mRNA | AK082119.1 | - | 1 | 1584 | ||
AK086765.1 | - | 1 | 1286 | |||
AK147476.1 | - | 2 | 7475 | |||
AK147682.1 | - | 2 | 7473 | |||
BC100546.1 | - | 1 | 1345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
