siRNA Details
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122952.1 | NP_001116424.1 | 11 | 9186 | ||
NM_001122953.1 | NP_001116425.1 | 10 | 9296 | |||
NM_010905.3 | NP_035035.1 | 11 | 9425 | |||
GenBank mRNA | AK034669.1 | - | 1 | 2794 | ||
AK053150.1 | - | 1 | 2494 | |||
AK173173.1 | - | 9 | 6111 | |||
BC094331.1 | - | 1 | 2196 |

More Information
Other Species Targeted

Gene Ontology Categories:
