siRNA Details
Gene Symbol: Vps13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177184.3 | NP_796158.2 | 4 | 284 | ||
GenBank mRNA | AB281675.1 | - | 4 | 284 | ||
AB281676.1 | - | 4 | 284 | |||
AB281677.1 | - | 4 | 284 | |||
AB281678.1 | - | 4 | 284 | |||
AB281681.1 | - | 1 | 66 | |||
AK045749.1 | - | 4 | 278 | |||
AK048766.1 | - | 4 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
