siRNA Details
Gene Symbol: PQBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032381.1 | NP_001027553.1 | 5 | 573 | ||
NM_001032382.1 | NP_001027554.1 | 5 | 641 | |||
NM_001032383.1 | NP_001027555.1 | 5 | 652 | |||
NM_001032384.1 | NP_001027556.1 | 5 | 561 | |||
NM_001167989.1 | NP_001161461.1 | 5 | 573 | |||
NM_001167990.1 | NP_001161462.1 | 5 | 531 | |||
NM_005710.2 | NP_005701.1 | 4 | 706 | |||
XM_005272571.3 | XP_005272628.1 | 5 | 636 | |||
XM_011543884.2 | XP_011542186.1 | 5 | 711 | |||
XM_017029207.1 | XP_016884696.1 | 5 | 702 | |||
GenBank mRNA | AB016533.1 | - | 5 | 622 | ||
AB041833.1 | - | 4 | 466 | |||
AB041834.1 | - | 4 | 452 | |||
AB041835.1 | - | 4 | 452 | |||
AJ005893.1 | - | 4 | 709 | |||
AJ242829.1 | - | 5 | 512 | |||
AJ973594.1 | - | 5 | 446 | |||
AJ973595.1 | - | 5 | 446 | |||
AJ973597.1 | - | 5 | 446 | |||
AJ973599.1 | - | 4 | 467 | |||
AJ973600.1 | - | 4 | 467 | |||
AJ973601.1 | - | 5 | 554 | |||
AJ973602.1 | - | 4 | 771 | |||
AJ973603.1 | - | 5 | 554 | |||
AJ973606.1 | - | 4 | 467 | |||
BC012358.1 | - | 5 | 533 | 4662649 | ||
BE385548.1 | - | 3 | 387 | |||
BE396796.1 | - | 5 | 553 | |||
BI818141.1 | - | 5 | 535 | |||
BX362311.2 | - | 5 | 610 | |||
CD051018.1 | - | 5 | 552 | |||
CN292761.1 | - | 5 | 641 | |||
KU178500.1 | - | 4 | 452 | |||
KU178501.1 | - | 5 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
