siRNA Details
Gene Symbol: Tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077418.1 | - | 2 | 352 | ||
AK080378.1 | - | 2 | 344 | |||
AK169483.1 | - | 2 | 344 | |||
BC057996.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
