siRNA Details
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 5 | 3272 | ||
GenBank mRNA | AK036482.1 | - | 1 | 483 | ||
AK043096.1 | - | 1 | 638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
