siRNA Details
Gene Symbol: Ncor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177229.3 | NP_796203.1 | 9 | 1187 | ||
GenBank mRNA | AK035813.1 | - | 9 | 1179 | ||
AK077420.1 | - | 9 | 1188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
