siRNA Details
Gene Symbol: Fsbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256142.1 | NP_001243071.1 | 1 | 289 | ||
GenBank mRNA | AK053419.1 | - | 1 | 288 | ||
AK147819.1 | - | 1 | 290 |

More Information
Other Species Targeted

Gene Ontology Categories:
