siRNA Details
Gene Symbol: Pisd-ps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132606.1 | - | 3 | 524 | ||
AK164215.1 | - | 4 | 550 |
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177298.3 | NP_796272.2 | 5 | 633 | ||
GenBank mRNA | AK033483.1 | - | 5 | 625 | ||
AK034656.1 | - | 5 | 635 | |||
AK162028.1 | - | 5 | 626 | |||
AK162737.1 | - | 5 | 634 | |||
AK167554.1 | - | 4 | 575 | |||
AK182065.1 | - | 2 | 125 | |||
BC070408.1 | - | 5 | 632 | 6809528 | ||
BC094594.1 | - | 5 | 624 | 5707749 | ||
BC116695.1 | - | 5 | 635 | 40105525 | ||
BC117509.1 | - | 5 | 635 | 40105529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
