siRNA Details
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177298.3 | NP_796272.2 | 8 | 1817 | ||
GenBank mRNA | AK033483.1 | - | 8 | 1809 | ||
AK034656.1 | - | 8 | 1819 | |||
AK162028.1 | - | 8 | 1810 | |||
AK167554.1 | - | 7 | 1759 | |||
BC070408.1 | - | 8 | 1816 | 6809528 | ||
BC094594.1 | - | 8 | 1808 | 5707749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
