siRNA Details
Gene Symbol: Rusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI849772.1 | - | 1 | 40 |
Gene Symbol: Fam166b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162381.1 | NP_001155853.1 | 5 | 1222 | ||
NM_177377.5 | NP_796351.2 | 4 | 1299 | |||
GenBank mRNA | AK029435.1 | - | 4 | 1300 | ||
AK053208.1 | - | 4 | 1300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
