siRNA Details
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177387.3 | NP_796361.2 | 8 | 1454 | ||
GenBank mRNA | AK029782.1 | - | 7 | 845 | ||
AK086181.1 | - | 8 | 1402 | |||
AK147850.1 | - | 8 | 1458 | |||
AK162950.1 | - | 8 | 1027 | |||
BC138155.1 | - | 8 | 1194 | 8861174 |
