siRNA Details
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177608.3 | NP_808276.2 | 2, 3 | 448 | ||
GenBank mRNA | AK049924.1 | - | 2, 3 | 449 | ||
AK145135.1 | - | 2, 3 | 333 | |||
AK169713.1 | - | 2, 3 | 335 | |||
AK172920.1 | - | 2, 3 | 317 | |||
BC095939.1 | - | 2, 3 | 315 | |||
BC141056.1 | - | 2, 3 | 408 | 9055668 |

More Information
Other Species Targeted

Gene Ontology Categories:
