siRNA Details
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081319.1 | NP_001074788.1 | 1, 2 | 394 | ||
GenBank mRNA | AK048394.1 | - | 1, 2 | 342 | ||
AK156653.1 | - | 1, 2 | 295 | |||
BC125448.1 | - | 1, 2 | 291 | |||
BC132346.1 | - | 1, 2 | 291 |

More Information
Other Species Targeted

Gene Ontology Categories:
