siRNA Details
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081319.1 | NP_001074788.1 | 2 | 427 | ||
GenBank mRNA | AK048394.1 | - | 2 | 375 | ||
AK156653.1 | - | 2 | 328 | |||
BC125448.1 | - | 2 | 324 | |||
BC132346.1 | - | 2 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
