siRNA Details
Gene Symbol: Igfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177642.3 | NP_808310.2 | 25 | 7932 | ||
GenBank mRNA | AK036509.1 | - | 6 | 707 | ||
BC107027.1 | - | 3 | 256 | 40050318 | ||
BC107028.2 | - | 3 | 256 | 40050319 | ||
EU429481.1 | - | 11 | 4804 | |||
EU429482.1 | - | 10 | 1671 | |||
EU429483.2 | - | 20 | 3085 | |||
GU290042.1 | - | 25 | 7932 |
