siRNA Details
Gene Symbol: Rasal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177644.5 | NP_808312.3 | 18 | 6706 | ||
GenBank mRNA | AK032643.1 | - | 1 | 2155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
