siRNA Details
Gene Symbol: Kansl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122738.1 | NP_001116210.1 | 9 | 2239 | ||
GenBank mRNA | AK082897.1 | - | 8 | 1059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
