siRNA Details
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005776.2 | NP_005767.1 | 3, 4 | 374 | ||
GenBank mRNA | AF070654.1 | - | 3, 4 | 327 | ||
AF104398.1 | - | 3, 4 | 327 | |||
AK074781.1 | - | 3, 4 | 279 | |||
AK311897.1 | - | 3, 4 | 287 | |||
AY358635.1 | - | 3, 4 | 284 | |||
BC103741.1 | - | 3, 4 | 375 | 6025795 | ||
BQ937490.1 | - | 3, 4 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
