siRNA Details
Gene Symbol: Cfap74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177674.5 | NP_808342.3 | 15, 16 | 2147 | ||
GenBank mRNA | AK040922.1 | - | 10, 11 | 1656 | ||
AK134858.1 | - | 15, 16 | 2079 | |||
BC116275.1 | - | 3, 4 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
