siRNA Details
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005787.5 | NP_005778.1 | 1, 2 | 227 | ||
GenBank mRNA | BC002839.2 | - | 1, 2 | 202 | 3636604 | |
BC004313.1 | - | 1, 2 | 182 | |||
BC021062.1 | - | 1, 2 | 191 | |||
KP189279.1 | - | 1, 2 | 220 | |||
KP189284.1 | - | 1, 2 | 218 | |||
KP189289.1 | - | 1, 2 | 217 | |||
KP189294.1 | - | 1, 2 | 196 | |||
KP189295.1 | - | 1, 2 | 196 | |||
KP189302.1 | - | 1, 2 | 217 | |||
KP189307.1 | - | 1, 2 | 216 | |||
KP189312.1 | - | 1, 2 | 216 | |||
KP189313.1 | - | 1, 2 | 216 | |||
KP189314.1 | - | 1, 2 | 216 | |||
Y09022.1 | - | 1, 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
