siRNA Details
Gene Symbol: Fat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183221.3 | NP_899044.3 | 1, 2 | 5203 | ||
GenBank mRNA | AK034552.1 | - | 1, 2 | 1201 | ||
DQ286572.1 | - | 1, 2 | 5203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
