siRNA Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039371.2 | NP_001034460.2 | 7, 8 | 1420 | ||
GenBank mRNA | AK029695.1 | - | 4, 5 | 578 | ||
BC094925.1 | - | 7, 8 | 1309 | |||
BC152347.1 | - | 2, 3 | 315 |

More Information
Other Species Targeted

Gene Ontology Categories:
