siRNA Details
Gene Symbol: Adgrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081342.1 | NP_001074811.1 | 22 | 2731 | ||
GenBank mRNA | AK054021.1 | - | 7 | 578 | ||
AK153762.1 | - | 22 | 2734 | |||
BC151161.1 | - | 22 | 2433 | 9088050 | ||
BC157994.1 | - | 21 | 2337 | 9007307 | ||
BC158001.1 | - | 21 | 2337 | 9007310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
