siRNA Details
Gene Symbol: Wdr66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030281.1 | - | 6 | 2880 | ||
BC138176.1 | - | 6 | 1012 | 8861195 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030281.1 | - | 6 | 2880 | ||
BC138176.1 | - | 6 | 1012 | 8861195 |