siRNA Details
Gene Symbol: 4933408B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177773.4 | NP_808441.2 | 3 | 1651 | ||
GenBank mRNA | AK030167.1 | - | 3 | 1651 | ||
BC172130.1 | - | 3 | 1525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
