siRNA Details
Gene Symbol: Prex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177782.3 | NP_808450.2 | 40 | 5270 | ||
GenBank mRNA | AK090301.1 | - | 14 | 1748 | ||
AK173168.1 | - | 40 | 5270 | |||
BC057617.1 | - | 15 | 1729 | |||
BC067047.1 | - | 38 | 5173 | |||
BC151074.1 | - | 40 | 5218 | 9087987 | ||
BC157940.1 | - | 40 | 5218 | 9088120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
