siRNA Details
Gene Symbol: Gm9975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050463.1 | - | 1 | 1251 | ||
BC042188.1 | - | 1 | 95 | |||
BC096608.1 | - | 1 | 1580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
